Läs mer
Di(N-desethyl) Amiodarone Hydrochloride, TRC
Produktkod. 30370427
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30370427 5 mg
1 options
Produktkod. 30370427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4407505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material di-N-desethyl Amiodarone hydrochloride
CAS 757220-04-5
Molekylformel C21 H21 I2 N O3 . Cl H
Kvantitet 5 mg
Synonym N,N-Di-Desethyl Amiodarone Hydrochloride
InChI formel InChI=1S/C21H21I2NO3.ClH/c1-2-3-7-18-19(14-6-4-5-8-17(14)27-18)20(25)13-11-15(22)21(16(23)12-13)26-10-9-24;/h4-6,8,11-12H,2-3,7,9-10,24H2,1H3;1H
LEDER Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN)c(I)c3
IUPAC-namn [4-(2-aminoethoxy)-3,5-diiodophenyl]-(2-butyl-1-benzofuran-3-yl)methanone;hydrochloride
Molekylvikt (g/mol) 625.67
Formel vikt 624.9378