Läs mer
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Produktkod. 11464314
Change view
Kvantitet:
10 g
50 g
250 g
Förpackningsstorlek:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11464314 250 g 250g
11444314 10 g 10g
11454314 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regleringsinformation: Denna produkt innehåller ett ämne som är upptaget i bilaga XIV till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.
Produktkod. 11464314 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A12593.30
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Regleringsinformation: Denna produkt innehåller ett ämne som är upptaget i bilaga XIV till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.
Solubility
Soluble in water (0.24 mg/L).
Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84-75-3
Molekylformel C20H30O4
Molekylvikt (g/mol) 334.46
MDL-nummer MFCD00043720
InChI-nyckel KCXZNSGUUQJJTR-UHFFFAOYSA-N
Synonym dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
PubChem CID 6786
ChEBI CHEBI:34678
IUPAC-namn dihexyl benzene-1,2-dicarboxylate
LEDER CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC
Specifikationer
Kemiskt namn eller material Di-n-hexyl phthalate
Densitet 1.01
Kokpunkt 185°C to 187°C (0.5 mmHg)
Brytningsindex 1.487
Kvantitet 250 g
UN-nummer UN3082
Löslighetsinformation Soluble in water (0.24mg/L).
Formel vikt 334.46
Procent renhet 97%