Läs mer
Di-n-hexyl Phthalate, TRC
Produktkod. 30179376
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179376 1 g
30179665 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4464901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84-75-3
Molekylformel C20 H30 O4
Molekylvikt (g/mol) 334.45
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dihexyl ester, 1,2-Benzenedicarboxylic acid, dihexyl ester (9CI), Phthalic acid, dihexyl ester (6CI,7CI,8CI), Bis(n-hexyl) phthalate, Di-n-hexyl phthalate, Dihexyl phthalate, Jayflex DHP, NSC 4817, Phthalic acid dihexyl ester, Phthalic acid, bis-hexyl ester
IUPAC-namn dihexyl benzene-1,2-dicarboxylate
LEDER CCCCCCOC(=O)c1ccccc1C(=O)OCCCCCC
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-hexyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14H,3-8,11-12,15-16H2,1-2H3
Formel vikt 334.2144
Procent renhet >95%