Läs mer
Di(p-anisyl)iodonium Bromide, TRC
Produktkod. 30349193
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30349193 5 g
1 options
Produktkod. 30349193 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4170605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Di(p-anisyl)iodonium Bromide
CAS 19231-06-2
Molekylformel C14H14BrIO2
Kvantitet 5 g
Synonym Bis(4-methoxyphenyl)iodonium Bromide, Bis(p-anisyl)iodonium Bromide, NSC 141350, p,p'-Dimethoxydiphenyliodonium Bromide
InChI formel InChI=1S/C14H14IO2.BrH/c1-16-13-7-3-11(4-8-13)15-12-5-9-14(17-2)10-6-12;/h3-10H,1-2H3;1H/q+1;/p-1
LEDER [Br-].COc1ccc([I+]c2ccc(OC)cc2)cc1
Molekylvikt (g/mol) 421.07
Formel vikt 419.9222