Läs mer
Di-p-tert-Butylphenyl Phosphorochloridate, TRC
Produktkod. 30377699
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30377699 500 mg
1 options
Produktkod. 30377699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD428535500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Di-p-tert-Butylphenyl Phosphorochloridate
CAS 38135-31-8
Molekylformel C20H26ClO3P
Kvantitet 500 mg
Synonym Bis[4-(1,1-dimethylethyl)phenyl] Ester Phosphorochloridic Acid
InChI formel InChI=1S/C20H26ClO3P/c1-19(2,3)15-7-11-17(12-8-15)23-25(21,22)24-18-13-9-16(10-14-18)20(4,5)6/h7-14H,1-6H3
LEDER O=P(OC1=CC=C(C(C)(C)C)C=C1)(Cl)OC2=CC=C(C(C)(C)C)C=C2
IUPAC-namn bis(4-(tert-butyl)phenyl) phosphorochloridate
Molekylvikt (g/mol) 380.85
Formel vikt 380.13