Läs mer
Di-p-tolyl-phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Di-p-tolyl-phosphate |
| CAS | 843-24-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C14H15O4P |
| Kvantitet | 25 mg |
| Synonym | Bis(4-methylphenyl) Ester Phosphoric Acid, Di-p-tolyl Ester Phosphoric Acid, p-Tolyl Phosphate ((C7H7O)2(HO)PO), Bis(4-methylphenyl) Phosphate, Bis(p-tolyl) phosphate, Di-p-tolyl Hydrogen Phosphate |
| InChI formel | InChI=1S/C14H15O4P/c1-11-3-7-13(8-4-11)17-19(15,16)18-14-9-5-12(2)6-10-14/h3-10H,1-2H3,(H,15,16) |
| LEDER | Cc1ccc(OP(=O)(O)Oc2ccc(C)cc2)cc1 |
| Molekylvikt (g/mol) | 278.24 |
| Formel vikt | 278.0708 |
| Visa mer |