Learn More
di-Potassium hydrogen phosphate trihydrate, Supelco™
Specifications
Specifications
| Chemical Name or Material | di-Potassium hydrogen phosphate trihydrate |
| Melting Point | 340°C (anhydrous substance; decomposes) |
| CAS | 16788-57-1 |
| Quantity | 25 kg |
| Synonym | di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic, di-Potassium hydrogen phosphate trihydrate |
| Molecular Weight (g/mol) | 228.22 |
| Grade | Not Available |