Läs mer
Di-sec-butyl Phthalate, TRC
Produktkod. 30369878
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30369878 1 g
1 options
Produktkod. 30369878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4295051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Di-sec-butyl Phthalate
CAS 4489-61-6
Molekylformel C16H22O4
Kvantitet 1 g
Synonym 1,2-Benzenedicarboxylic Acid 1,2-Bis(1-methylpropyl) Ester, Phthalic Acid Di-sec-butyl Ester
InChI formel InChI=1S/C16H22O4/c1-5-11(3)19-15(17)13-9-7-8-10-14(13)16(18)20-12(4)6-2/h7-12H,5-6H2,1-4H3
LEDER CCC(C)OC(=O)c1ccccc1C(=O)OC(C)CC
Molekylvikt (g/mol) 278.34
Formel vikt 278.1518