Learn More
di-Sodium hydrogen phosphate dihydrate, MilliporeSigma™
Product Code. 15874870
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
Unit Size:
500g
Product Code. Quantity Packaging Unit Size
15874870 500 g Plastic Bottle 500g
1 options
Product Code. 15874870 Supplier Supelco Supplier No. 1065800500
 Restricted Item
Beräknad leverans: 19-11-2026
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 92.5°C
CAS 10028-24-7
pH 9 to 9.4
Physical Form Solid
Quantity 500 g
Molecular Formula H5Na2O6P
MDL Number MFCD00149182
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium dihydrate hydrogen phosphate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99
Percent Purity ≥99.5%
Grade Not Available
Packaging Plastic Bottle
Density 2.1g/cm3 (20°C)
Chemical Name or Material di-Sodium hydrogen phosphate dihydrate