missing translation for 'onlineSavingsMsg'
Läs mer

Di-tert-butyl N,N-diethylphosphoramidite, 94%

Produktkod. p-7029867
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11367169 1 g 1g
11377169 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11367169 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27380.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 117924-33-1 | C12H28NO2P | 249.335 g/mol

A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

Solubility
Hydrolyzes in water. Soluble in chloroform and methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 117924-33-1
Molekylformel C12H28NO2P
Molekylvikt (g/mol) 249.335
MDL-nummer MFCD00134307
InChI-nyckel KUKSUQKELVOKBH-UHFFFAOYSA-N
Synonym di-tert-butyl n,n-diethylphosphoramidite, di-tert-butyl diethylphosphoramidite, di-t-butyl n,n-diethylphosphoramidite, phosphoramidous acid, diethyl-, bis 1,1-dimethylethyl ester, n-bis 2-methylpropan-2-yl oxy phosphanyl-n-ethylethanamine, acmc-1c42i, di-t-butyl diethylphosphoramidite, di-tert-butyl diethylamidophosphite, di-tert-butoxy diethylamino phosphine
PubChem CID 4251849
IUPAC-namn N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-ethylethanamine
LEDER CCN(CC)P(OC(C)(C)C)OC(C)(C)C

Specifikationer

Kemiskt namn eller material Di-tert-butyl N,N-diethylphosphoramidite
Densitet 0.896
Kokpunkt 39°C to 41°C (0.3 mmHg)
Flampunkt 78°C (172°F)
Brytningsindex 1.434
Kvantitet 1 g
Känslighet Moisture sensitive; May form a precipitate
Löslighetsinformation Hydrolyzes in water. Soluble in chloroform and methanol.
Formel vikt 249.33
Procent renhet 94%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.