Läs mer
Diacetyl Bis(2,4-dinitrophenylhydrazone), TRC
Produktkod. 30134650
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134650 2.5 g
30006886 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4225302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1179-29-9
Molekylformel C16H14N8O8
Molekylvikt (g/mol) 446.33
Synonym 2,3-Bis[2-(2,4-dinitrophenyl)hydrazone] 2,3-Butanedione, Bis[(2,4-dinitrophenyl)hydrazone] 2,3-Butanedion, NSC 409622;
IUPAC-namn N-[3-[(2,4-dinitrophenyl)hydrazinylidene]butan-2-ylideneamino]-2,4-dinitroaniline
LEDER CC(=NNC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C(=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-])C
Specifikationer
Kemiskt namn eller material Diacetyl Bis(2,4-dinitrophenylhydrazone)
Kvantitet 2.5 g
InChI formel InChI=1S/C16H14N8O8/c1-9(17-19-13-5-3-11(21(25)26)7-15(13)23(29)30)10(2)18-20-14-6-4-12(22(27)28)8-16(14)24(31)32/h3-8,19-20H,1-2H3
Formel vikt 446.0935