Learn More
Diaion™ PA408(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type II, 0.9 meq/ml on PS-DVB, Thermo Scientific™
Sugar refining/decolorizing, particularly beet sugar
Brand: Thermo Scientific Alfa Aesar 046474.A1
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
300-1180μm Beads | |
4% cross-linked, strongly basic porous type II, 0.9meq/mL on PS-DVB | |
300 to 1180μm |
Diaion™ PA408 (Cl), ion exchange resin | |
Amine-like | |
1 kg |