Learn More
Diaion™ PK220(Na), ion exchange resin, 10% cross-linked, strongly acidic porous type, 1.9 meq/ml on PS-DVB, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar 046511.A1
Additional Details : Weight : 1.00000kg
Product Code. 15494045
  • 1114.74 SEK / 1kg
Estimated Shipment: 27-05-2024
to see stock.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
  • Used in solutions of high electrolyte content
Specifications
Specifications
300-1180μm Beads
10% cross-linked, strongly acidic porous type, 1.9meq/mL on PS-DVB
1 kg
Diaion™ PK220 (Na), ion exchange resin
300 to 1180μm
Product Suggestions
Product Suggestions
Videos
SDS Documents