Learn More
Diaion™ SA11A(Cl), ion exchange resin, lightly cross-linked, strongly basic gel type I, 0.85 meq/ml on PS-DVB, Thermo Scientific™
Decolorization in sugar solutions, metal recovery, pharmaceuticals
Brand: Thermo Scientific Alfa Aesar 046980.A1
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
300-1180μm Beads | |
Lightly cross-linked, strongly basic gel type I, 0.85meq/mL on PS-DVB | |
300 to 1180μm |
Diaion™ SA11A (Cl), ion exchange resin | |
Odorless | |
1 kg |