Läs mer
Dialkyl Phthalate (Alkyl= Heptyl:Heptyl, Nonyl:Nonyl, Undecyl:Undecyl, Heptyl:Nonyl, Heptyl:Undecyl, Nonyl:Undecyl), TRC
Produktkod. 30179458
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179458 1 g
30178942 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179458 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4149001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68515-42-4
Molekylformel C8 H4 O4 . 2(Cx H2x+1)
Molekylvikt (g/mol) 418.6
Synonym Dialkyl Phthalate (Alkyl= Heptyl:Heptyl, Nonyl:Nonyl, Undecyl:Undecyl, Heptyl:Nonyl, Heptyl:Undecyl, Nonyl:Undecyl)
IUPAC-namn heptyl undecyl phthalate
LEDER [CH2+1]OC(C1=CC=CC=C1C(O[CH2+1])=O)=O
Specifikationer
Kemiskt namn eller material Dialkyl Phthalate
Kvantitet 1 g
InChI formel InChI=1S/C26H42O4/c1-3-5-7-9-10-11-12-14-18-22-30-26(28)24-20-16-15-19-23(24)25(27)29-21-17-13-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3