Läs mer
Dialkyl Phthalate (Alkyl= Heptyl:Heptyl, Nonyl:Nonyl, Undecyl:Undecyl, Heptyl:Nonyl, Heptyl:Undecyl,, TRC
Produktkod. 30368511
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30368511 100 mg
1 options
Produktkod. 30368511 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD414900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dialkyl Phthalate
CAS 68515-42-4
Molekylformel C8 H4 O4 . 2(Cx H2x+1)
Kvantitet 100 mg
Synonym Dialkyl Phthalate (Alkyl= Heptyl:Heptyl, Nonyl:Nonyl, Undecyl:Undecyl, Heptyl:Nonyl, Heptyl:Undecyl, Nonyl:Undecyl)
InChI formel InChI=1S/C26H42O4/c1-3-5-7-9-10-11-12-14-18-22-30-26(28)24-20-16-15-19-23(24)25(27)29-21-17-13-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3
LEDER [CH2+1]OC(C1=CC=CC=C1C(O[CH2+1])=O)=O
IUPAC-namn heptyl undecyl phthalate
Molekylvikt (g/mol) 418.6