Läs mer
Diallyl Tetrasulfide (>85%), TRC
Produktkod. 30353369
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30353369 25 mg
1 options
Produktkod. 30353369 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41598525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diallyl Tetrasulfide
CAS 2444-49-7
Molekylformel C6 H10 S4
Kvantitet 25 mg
Synonym Diallyl Tetrasulfide
InChI formel InChI=1S/C6H10S4/c1-3-5-7-9-10-8-6-4-2/h3-4H,1-2,5-6H2
LEDER C=CCSSSSCC=C
IUPAC-namn 3-(prop-2-enyltetrasulfanyl)prop-1-ene
Molekylvikt (g/mol) 210.404
Formel vikt 209.967