Läs mer
Diallylacetic Acid Ethyl Ester, TRC
Produktkod. 30362118
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30362118 100 mg
1 options
Produktkod. 30362118 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD415995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diallylacetic Acid Ethyl Ester
CAS 18325-74-1
Molekylformel C10H16O2
Kvantitet 100 mg
Synonym 2-(2-Propenyl)-4-pentenoic Acid Ethyl Ester, 2-Allyl-4-pentenoic Acid Ethyl Ester, Ethyl 2-(2-Propenyl)-4-pentenoate, Ethyl 2-(Prop-2'-enyl)-4-pentenoate, Ethyl 2-Allyl-4-pentenoate
InChI formel InChI=1S/C10H16O2/c1-4-7-9(8-5-2)10(11)12-6-3/h4-5,9H,1-2,6-8H2,3H3
LEDER C=CCC(CC=C)C(OCC)=O
IUPAC-namn ethyl 2-allylpent-4-enoate
Molekylvikt (g/mol) 168.23
Formel vikt 168.12