Läs mer
Diallylmalonic Acid, TRC
Produktkod. 30356831
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30356831 250 mg
1 options
Produktkod. 30356831 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD415970250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diallylmalonic Acid
CAS 4372-31-0
Molekylformel C9H12O4
Kvantitet 250 mg
Synonym alpha,alpha-Diallylmalonic Acid, 1,6-Heptadiene-4,4-dicarboxylic Acid, Di-2-propenyl-propanedioic Acid, 2,2-Di-2-propen-1-ylpropanedioic Acid
InChI formel InChI=1S/C9H12O4/c1-3-5-9(6-4-2,7(10)11)8(12)13/h3-4H,1-2,5-6H2,(H,10,11)(H,12,13)
LEDER OC(=O)C(CC=C)(CC=C)C(=O)O
Molekylvikt (g/mol) 184.19
Formel vikt 184.0736