Läs mer
Diamidafos, TRC
Produktkod. 30359773
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30359773 100 mg
1 options
Produktkod. 30359773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD120100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diamidafos
CAS 1754-58-1
Molekylformel C8 H13 N2 O2 P
Kvantitet 100 mg
Synonym Diamidafos, Diamidaphos, Diamidfos, Dowco 169, Nellite, O-Phenyl-N,N′-dimethyl-phosphorodiamidate
InChI formel InChI=1S/C8H13N2O2P/c1-9-13(11,10-2)12-8-6-4-3-5-7-8/h3-7H,1-2H3,(H2,9,10,11)
LEDER CNP(=O)(NC)Oc1ccccc1
IUPAC-namn N-[methylamino(phenoxy)phosphoryl]methanamine
Molekylvikt (g/mol) 200.175
Formel vikt 200.071