missing translation for 'onlineSavingsMsg'
Läs mer

Dianhydro Idarubicinaglycone, TRC

Produktkod. 30360646
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360646 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30360646 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3030455MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Dianhydro Idarubicinaglycone
CAS 84499-12-7
Molekylformel C20 H12 O5
Kvantitet 5 mg
Synonym 8-Acetyl-6,11-Dihydroxytetracene-5,12-dione, Dianhydro Idarubicinaglycone
InChI formel InChI=1S/C20H12O5/c1-9(21)10-6-7-13-14(8-10)20(25)16-15(19(13)24)17(22)11-4-2-3-5-12(11)18(16)23/h2-8,24-25H,1H3
LEDER CC(=O)c1ccc2c(O)c3C(=O)c4ccccc4C(=O)c3c(O)c2c1
IUPAC-namn 8-acetyl-6,11-dihydroxytetracene-5,12-dione
Molekylvikt (g/mol) 332.306
Formel vikt 332.068
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.