Läs mer
Diaquo[(1R,2R)-1,2-cyclohexanediamine]platinum Dimer Dinitrate, TRC
Produktkod. 17994985
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994985 10 mg
17931275 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17994985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41676510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82398-34-3
Molekylformel C12H30N6O8Pt2
Molekylvikt (g/mol) 776.56
Synonym (SP-4-2)-Di-oxobis[(1R,2R)-1,2-cyclohexanediamine-κN,κN']diplatinum(2+) Dinitrate, (SP-4-2)-Di-oxobis[(1R-trans)-1,2-cyclohexanediamine-N,N']diplatinum(2+) Dinitrate;
LEDER [H][O-]1[Pt+2]2([O-]([H])[Pt+2]31[NH2][C@@H]4CCCC[C@H]4[NH2]3)[NH2][C@@H]5CCCC[C@H]5[NH2]2
Specifikationer
Kemiskt namn eller material Diaquo[(1R,2R)-1,2-cyclohexanediamine]platinum Dimer Dinitrate
Kvantitet 10 mg
InChI formel InChI=1S/2C6H14N2.2H2O.2Pt/c2*7-5-3-1-2-4-6(5)8;;;;/h2*5-6H,1-4,7-8H2;2*1H2;;/q;;;;2*+4/p-2/t2*5-,6-;;;;/m11..../s1