Läs mer
Diaquo[(1R,2R)-1,2-cyclohexanediamine]platinum Dinitrate, TRC
Produktkod. 30356668
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30356668 10 mg
1 options
Produktkod. 30356668 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41676010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diaquo[(1R,2R)-1,2-cyclohexanediamine]platinum Dinitrate
CAS 94042-08-7
Molekylformel C6 H18 N2 O2 Pt . 2 N O3
Kvantitet 10 mg
Synonym Platinum(2+), diaqua[(1R,2R)-1,2-cyclohexanediamine-κN1,κN2]-, (SP-4-2)-, nitrate (1:2), Platinum(2+), diaqua(1,2-cyclohexanediamine-N,N')-, [SP-4-2-(1R-trans)]-, dinitrate, Platinum(2+), diaqua[(1R,2R)-1,2-cyclohexanediamine-κN,κN']-, (SP-4-2)-, dinitrate (9CI), 1,2-Cyclohexanediamine, platinum complex, (1R-trans)-, Diaqua(1R,2R-cyclohexanediammine)platinum(II) dinitrate, Diaquo[trans-(-)-1,2-cyclohexanediamine]platinum(II) dinitrate, Platinum(2+), diaqua(1,2-cyclohexanediamine-κN,κN')-, [SP-4-2-(1R-trans)]-, dinitrate, (R,R)-Diaquodiaminocyclohexaneplatinum Dinitrate
InChI formel InChI=1S/C6H14N2.2NO3.2H2O.Pt/c7-5-3-1-2-4-6(5)8;2*2-1(3)4;;;/h5-6H,1-4,7-8H2;;;2*1H2;/q;2*-1;;;+2/t5-,6-;;;;;/m1...../s1
LEDER [O-][N+](=O)[O-].[O-][N+](=O)[O-].C1CC[C@@H]2[C@@H](C1)[NH2][Pt+2]([NH2]2)([OH2])[OH2]
Molekylvikt (g/mol) 469.31
Formel vikt 469.0772