Läs mer
Diazinon, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diazinon |
| CAS | 333-41-5 |
| Molekylformel | C12 H21 N2 O3 P S |
| Kvantitet | 500 mg |
| Synonym | Phosphorothioic acid, O,O-diethyl 2-isopropyl-6-methyl-4-pyrimidinyl ester (6CI), Phosphorothioic acid, O,O-diethyl O-(2-isopropyl-6-methyl-4-pyrimidinyl) ester (8CI), Antigal, Bassadinon, Basudin, Basudin 10G, Basudin 5G, Basudin 600 EW, Basudin S, Bazanon, Ciazinon, Compass, Compass (insecticide), Cooper's Flystrike Powder, Dacutox, Dassitox, Dazzel, Delzinon, Dew 600EW, Diazinon, Diazinon 4E, Diazinon AG 500, Diazinon G, Dicid, Diethyl 2-isopropyl-4-methyl-6-pyrimidinyl phosphorothionate, Diethyl 2-isopropyl-4-methyl-6-pyrimidyl thionophosphate, Dimpylat, Dimpylate, Disonex, Dizictol, Dizinil, Drawizon, ENT 19,507, ENT 19507, Evict, Exodin, Flytrol, G 24480, G 301, Galesan, Garden Tox, Hi-Yield Imported Fire Ant Killer, KFM Blowfly Dressing, Kleen-Dok, Knox Out 2FM, Knox-out, Meodinon, NSC 8938, Nasr-Cidol, Neocidol, Neocidol (oil), O,O-Diethyl O-(2-isopropyl-4-methyl-6-pyrimidinyl) phosphorothioate, O,O-Diethyl O-(2-isopropyl-4-methyl-6-pyrimidyl) thionophosphate, O,O-Diethyl O-(2-isopropyl-6-methyl-4-pyrimidinyl) phosphorothioate, O,O-Diethyl-O-(2-isopropyl-4-methyl-6-pyrimidyl) thiophosphate, Oleodiazinon, Optimizer, Ortho Diazinon Ultra, Sarolex, Spectracide, Spectracide 25EC, Terminator, Topclip 40, Topclip Blue, Topclip Blue Shield |
| InChI formel | InChI=1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3 |
| LEDER | CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)C |
| IUPAC-namn | diethoxy-(6-methyl-2-propan-2-ylpyrimidin-4-yl)oxy-sulfanylidene-λ^{5}-phosphane |
| Molekylvikt (g/mol) | 304.35 |
| Formel vikt | 304.101 |