missing translation for 'onlineSavingsMsg'
Läs mer

Dibenzo(A,L)Pyrene, SPEX CertiPrep™

Produktkod. 12969154 Handla allt SPEX Certiprep Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mL
Förpackningsstorlek:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
12969154 1 mL 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 12969154 Leverantör SPEX Certiprep Leverantörsnummer S1196

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Innehåller

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Dibenzo(a,l)pyrene191-30-01000μg/mL

Certifikat

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Uppfyllande

GHS/OSHA Complaint

Specifications

Kemiskt namn eller material Dibenzo(a,l)pyrene
Förpackning 2mL Amber Prescored Glass Ampule
CAS 191-30-0, 75-09-2
CAS Min % 0.1, 99.9
Anteckningar om koncentration eller sammansättning Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylformel C24H14
För användning med (utrustning) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI-nyckel JNTHRSHGARDABO-UHFFFAOYSA-N
LEDER C1=CC=C2C(=C1)C=C3C=CC4=C5C3=C2C6=CC=CC=C6C5=CC=C4
Lösningsmedel eller matris MeCl
Molekylvikt (g/mol) 302.376
PubChem CID 9119
Kvantitet 1 mL
ChEBI CHEBI:35861
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.