missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[B,K]fluoranthene, TRC

Product Code. 30378259
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30378259 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378259 Supplier Toronto Research Chemicals Supplier No. TRCD48574025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Dibenzo(b,k)fluoranthene
CAS 205-97-0
Molekylformel C24 H14
Kvantitet 25 mg
InChI formel InChI=1S/C24H14/c1-2-7-16-13-22-21(12-15(16)6-1)20-11-5-10-19-18-9-4-3-8-17(18)14-23(22)24(19)20/h1-14H
LEDER c1ccc2cc3c(cc2c1)c4cccc5c6ccccc6cc3c45
Molekylvikt (g/mol) 302.37
Formel vikt 302.1096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.