Läs mer
Dibromoformaldoxime, TRC
Produktkod. 30375773
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30375773 1 g
1 options
Produktkod. 30375773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4253001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dibromoformaldoxime
CAS 74213-24-4
Molekylformel CHBr2NO
Kvantitet 1 g
Synonym Hydroxy-carbonimidic Dibromide, Dibromoformaldehyde Oxime
InChI formel InChI=1S/CHBr2NO/c2-1(3)4-5/h5H
LEDER ON=C(Br)Br
IUPAC-namn N-(dibromomethylidene)hydroxylamine
Molekylvikt (g/mol) 202.83
Formel vikt 200.8425