missing translation for 'onlineSavingsMsg'
Läs mer

Dibutyl 3,3'-((2-Hydroxyethyl)azanediyl)dipropionate, TRC

Produktkod. 30366663
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366663 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30366663 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD224950100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Dibutyl 3,3'-((2-Hydroxyethyl)azanediyl)dipropionate
CAS 1372787-88-6
Molekylformel C16H31NO5
Kvantitet 100 mg
Synonym N-(3-Butoxy-3-oxopropyl)-N-(2-hydroxyethyl)-b-alanine butyl ester
InChI formel InChI=1S/C16H31NO5/c1-3-5-13-21-15(19)7-9-17(11-12-18)10-8-16(20)22-14-6-4-2/h18H,3-14H2,1-2H3
LEDER O=C(OCCCC)CCN(CCO)CCC(=O)OCCCC
IUPAC-namn butyl 3-[(3-butoxy-3-oxopropyl)-(2-hydroxyethyl)amino]propanoate
Molekylvikt (g/mol) 317.43
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.