missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Dibutyl Phosphate-d18, TRC
Specifications
Specifications
| Kemiskt namn eller material | Dibutyl Phosphate-d18 |
| CAS | 156213-21-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C8HD18O4P |
| Kvantitet | 50 mg |
| Synonym | 1-Butan-1,1,2,2,3,3,4,4,4-d18-ol Hydrogen Phosphate, Butyl Phosphate, (BuO)2(HO)PO-d18, DBP-d18, DP 4-d18, DP 4 (coupling agent)-d18, Di-n-Butyl Phosphate-d18, Dibutyl Acid Phosphate-d18, Dibutyl Hydrogen Phosphate-d18, Dibutyl Phosphate-d18, HDBP-d18, NSC 203039-d18, NSC 22313-d18, Phosphoric Acid Dibutyl Ester-d18 |
| InChI formel | InChI=1S/C8H19O4P/c1-3-5-7-11-13(9,10)12-8-6-4-2/h3-8H2,1-2H3,(H,9,10)/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2 |
| LEDER | O=P(OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H] |
| Molekylvikt (g/mol) | 228.32 |
| Procent renhet | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?