Läs mer
Dibutylammonium Acetate (0.5M Solution in Water), TRC
Produktkod. 30132907
Change view
Kvantitet:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132907 500 mL
30012245 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132907 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD427250500ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19070-91-8
Molekylformel C8H19N . C2H4O2
Molekylvikt (g/mol) 189.3
Synonym N-Butyl-1-butanamine Acetate, Dibutylamine Acetate, Acetic Acid Dibutylamine Salt
IUPAC-namn acetic acid;N-butylbutan-1-amine
LEDER CCCCNCCCC.CC(=O)O
Specifikationer
Kemiskt namn eller material Dibutylammonium Acetate
Kvantitet 500 mL
InChI formel InChI=1S/C8H19N.C2H4O2/c1-3-5-7-9-8-6-4-2;1-2(3)4/h9H,3-8H2,1-2H3;1H3,(H,3,4)
Formel vikt 189.1729
Koncentration eller sammansättning (efter analyt eller komponenter) Dibutylammonium Acetate 0.5M