Läs mer
Dicamba, TRC
Produktkod. 30377884
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30377884 100 mg
1 options
Produktkod. 30377884 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD429800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dicamba
CAS 1918-00-9
Molekylformel C8 H6 Cl2 O3
Kvantitet 100 mg
Synonym o-Anisic acid, 3,6-dichloro- (8CI), 3,6-Dichloro-2-methoxybenzoic acid, 2,5-Dichloro-6-methoxybenzoic acid, 2-Methoxy-3,6-dichlorobenzoic acid, 3,6-Dichloro-o-anisic acid, Aviator, Banvel, Banvel 480, Banvel 70WP, Banvel SGF, Celebrity B, DC, Dianat, Dicamba, MDBA, Mais Banvel WG, Mediben, Vanquish, Velsicol 58CS11, Velsicol Compound R
InChI formel InChI=1S/C8H6Cl2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3H,1H3,(H,11,12)
LEDER COc1c(Cl)ccc(Cl)c1C(=O)O
IUPAC-namn 3,6-dichloro-2-methoxybenzoic acid
Molekylvikt (g/mol) 221.04
Formel vikt 219.9694