Learn More
Alfa Aesar™ Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%
Coupling reactions, Suzuki couplings
Brand:  Alfa Aesar™ 044829.01
Additional Details : CAS Number : 95408-45-0 Weight : 0.00025kg
Quantity 0.25g
 View more versions of this product
Product Code. 15462037
  • 891.40 SEK / 0.25g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)
C26H44Cl2FeP2Pd
MFCD08064219
95408-45-0
651.77
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
Specifications
Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)
Crystalline
651.77
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
Pd 16.3%
95408-45-0
C26H44Cl2FeP2Pd
MFCD08064219
Dark red
0.25g