Läs mer
Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer |
| CAS | 12354-85-7 |
| Molekylformel | 2 C10 H16 . 4 Cl . 2 Rh |
| Kvantitet | 50 mg |
| Synonym | 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene Rhodium Complex, (eta5-Pentamethylcyclopentadienyl)rhodium Dichloride Dimer, Bis(chloro(μ-chloro)(eta5-pentamethylcyclopentadienyl)rhodium), Bis(dichloro(pentamethylcyclopentadienyl)rhodium), Bis(pentamethylcyclopentadienylrhodium dichloride), Bis(μ-chloro)bis(chloro(eta5-pentamethylcyclopentadienyl)rhodium), Bis[dichloro[eta5-(pentamethylcyclopentadienyl)]rhodium], Di-μ-Chlorodichlorobis(pentamethyl-π-cyclopentadienyl)dirhodium, Di-μ-chlorobis[chloro(eta5-pentamethylcyclopentadienyl)rhodium], Di-μ-chlorodichlorobis((eta5-pentamethylcyclopentadienyl)rhodium), Di-μ-chlorodichlorobis(pentamethylcyclopentadienyl)dirhodium, Dichloro(pentamethylcyclopentadienyl)rhodium Dimer, Dichlorobis(μ2-chloro)bis(eta5-pentamethylcyclopentadienyl)dirhodium, Pentamethylcyclopentadienerhodium Dichloride Dimer, Tetrachlorobis(pentamethylcyclopentadienyl)dirhodium, etrachlorobis(eta5-pentamethylcyclopentadienyl)dirhodium, Di-μ-chlorodichlorobis[(1,2,3,4,5-eta)-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl]di-rhodium |
| InChI formel | InChI=1S/2C10H16.4ClH.2Rh/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*6H,1-5H3;4*1H;;/q2*-1;;;;;2*+3/p-4 |
| LEDER | [Cl-].[Cl-].[Cl-].[Cl-].[Rh+3].[Rh+3].CC1[C-](C)[C](C)C(=C1C)C.CC2[C-](C)[C](C)C(=C2C)C |
| IUPAC-namn | 1,2,3,4,5-pentamethylcyclopentene;rhodium(3+);tetrachloride |
| Molekylvikt (g/mol) | 620.091 |
| Formel vikt | 617.937 |