Learn More
Diclazuril-methyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30316377
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30316377 1 mL 1mL
1 options
Product Code. 30316377 Supplier Dr. Ehrenstorfer Supplier No. DREA12533200AL100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diclazuril-methyl
CAS 103337-71-9
Analyte or Component Names Diclazuril-methyl
Shipping Condition Room Temperature
Molecular Formula C18 H11 Cl3 N4 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzeneacetonitrile, 2,6-dichloro-α-(4-chlorophenyl)-4-(4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl)-α-methyl-, 2,6-Dichloro-α-(4-chlorophenyl)-4-(4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl)-α-methylbenzeneacetonitrile, Diclazuril-methyl
InChI Formula InChI=1S/C18H11Cl3N4O2/c1-18(9-22,10-2-4-11(19)5-3-10)16-13(20)6-12(7-14(16)21)25-17(27)24-15(26)8-23-25/h2-8H,1H3,(H,24,26,27)
SMILES CC(C#N)(c1ccc(Cl)cc1)c2c(Cl)cc(cc2Cl)N3N=CC(=O)NC3=O
IUPAC Name 2-(4-chlorophenyl)-2-[2,6-dichloro-4-(3,5-dioxo-1,2,4-triazin-2-yl)phenyl]propanenitrile
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 421.66
Quantity 1 mL
Formula Weight 419.9948 g/mol