Läs mer
Diclofenac-13C6, TRC
Produktkod. 30005236
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005236 5 mg
30008304 10 mg
30007717 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4364495MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1261393-71-8
Molekylformel C813C6H11Cl2NO2
Molekylvikt (g/mol) 302.1
Synonym 2-[(2,6-Dichlorophenyl)amino]-benzene-1,2,3,4,5,6-13C6-Acetic Acid
LEDER ClC1=CC=CC(Cl)=C1N[13C]2=[13C](CC(O)=O)[13C]([H])=[13C]([H])[13C]([H])=[13C]2[H]
Specifikationer
Kemiskt namn eller material Diclofenac-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C14H11Cl2NO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17H,8H2,(H,18,19)/i1+1,2+1,4+1,7+1,9+1,12+1
Formel vikt 301.0368
Procent renhet >95%