Läs mer
Diclofenac Dimer Impurity, TRC
Produktkod. 17995075
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17995075 2.5 mg
17980915 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17995075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4364552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1609187-30-5
Molekylformel C28H20Cl4N2O4
Molekylvikt (g/mol) 590.28
LEDER ClC(C=CC=C1Cl)=C1NC2=C(CC(O)=O)C=C(C3=CC=C(NC4=C(Cl)C=CC=C4Cl)C(CC(O)=O)=C3)C=C2
Specifikationer
Kemiskt namn eller material Diclofenac Dimer Impurity
Kvantitet 2.5 mg
InChI formel InChI=1S/C28H20Cl4N2O4/c29-19-3-1-4-20(30)27(19)33-23-9-7-15(11-17(23)13-25(35)36)16-8-10-24(18(12-16)14-26(37)38)34-28-21(31)5-2-6-22(28)32/h1-12,33-34H,13-14H2,(H,35,36)(H,37,38)