Läs mer
Diclofenac Ethyl Ester, TRC
Produktkod. 30003435
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003435 50 mg
30011911 250 mg
30009918 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43649550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15307-77-4
Molekylformel C16 H15 Cl2 N O2
Molekylvikt (g/mol) 324.2
Synonym Ethyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Ethyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Aceclofenac Imp. C (EP), Ethyl Ester of Diclofenac
IUPAC-namn ethyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
LEDER CCOC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
Specifikationer
Kemiskt namn eller material Diclofenac Ethyl Ester
Kvantitet 50 mg
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C16H15Cl2NO2/c1-2-21-15(20)10-11-6-3-4-9-14(11)19-16-12(17)7-5-8-13(16)18/h3-9,19H,2,10H2,1H3
Formel vikt 323.048
Procent renhet >95%