Läs mer
Diclofenac Isopropyl Ester, TRC
Produktkod. 30003639
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003639 25 mg
30009127 100 mg
30012855 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43645825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66370-79-4
Molekylformel C17 H17 Cl2 N O2
Molekylvikt (g/mol) 338.23
Synonym Isopropyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Isopropyl Ester of Diclofenac
IUPAC-namn propan-2-yl 2-[2-(2,6-dichloroanilino)phenyl]acetate
LEDER CC(C)OC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
Specifikationer
Kemiskt namn eller material Isopropyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Isopropyl Ester of Diclofenac)
Kvantitet 25 mg
InChI formel InChI=1S/C17H17Cl2NO2/c1-11(2)22-16(21)10-12-6-3-4-9-15(12)20-17-13(18)7-5-8-14(17)19/h3-9,11,20H,10H2,1-2H3
Formel vikt 337.0636