Läs mer
Diclofenac Potassium Salt, TRC
Produktkod. 30372939
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30372939 250 mg
1 options
Produktkod. 30372939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD436460250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diclofenac potassium
CAS 15307-81-0
Molekylformel C14 H10 Cl2 N O2 . K
Kvantitet 250 mg
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, potassium salt (1:1), Acetic acid, [o-(2,6-dichloroanilino)phenyl]-, monopotassium salt (8CI), Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, monopotassium salt (9CI), CGP 45840B, Caflam, Cambia, Cataflam, Diclofenac potassium, K-Fenak, K-fenak, Potassium diclofenac, ProSorb, Zipsor
InChI formel InChI=1S/C14H11Cl2NO2.K/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1
LEDER [K+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC-namn potassium;2-[2-(2,6-dichloroanilino)phenyl]acetate
Molekylvikt (g/mol) 334.24
Formel vikt 332.9726