Läs mer
Diclofensine hydrochloride, MedChemExpress
Produktkod. 16433557
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16433557 50 mg
16420018 5 mg
16417538 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16433557 Leverantör Medchem Express Leverantörsnummer HY18610
MedChemExpress Diclofensine hydrochloride (Ro-8-4650 hydrochloride) is a potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively.
Diclofensine hydrochloride (Ro-8-4650 hydrochloride) is a potent inhibitor of monoamine reuptake, blocking the uptake of dopamine, noradrenaline, and serotonin by rat brain synaptosomes with IC50 values of 0.74, 2.3, and 3.7 nM, respectively.
Specifikationer
Kemiskt namn eller material Diclofensine hydrochloride
CAS 34041-84-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C17H18Cl3NO
Kvantitet 50 mg
Synonym Ro 8-4650 hydrochloride
Löslighetsinformation H2O : ≥ 52 mg/mL (144.97 mM)
LEDER CN1CC2=C(C=CC(OC)=C2)C(C3=CC=C(Cl)C(Cl)=C3)C1.[H]Cl
Molekylvikt (g/mol) 358.69
Formel vikt 358.69
Procent renhet 99.88%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation