Learn More
Dicyandiamide, 99%, Thermo Scientific Chemicals
Catalog Number p-7080772
Click to view available options
Quantity:
250 g
500 g
2500 g
461-58-5
C2H4N4
84.08
MFCD00008066
QGBSISYHAICWAH-UHFFFAOYSA-N
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
10005
2-cyanoguanidine
NC(N)=NC#N
461-58-5
C2H4N4
84.08
MFCD00008066
QGBSISYHAICWAH-UHFFFAOYSA-N
dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
10005
2-cyanoguanidine
NC(N)=NC#N
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.
Solubility
Soluble in Water (32g/L ) at 20°C
Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
TRUSTED_SUSTAINABILITY
Melting Point 208°C to 211°C
Density 1.4
Assay Percent Range 99%
Odor Odorless
Quantity 250 g
Beilstein 605637
Merck Index 14,3092
Solubility Information Soluble in Water (32g/L ) at 20°C
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material Dicyandiamide