Läs mer
Dicyclohexylmethanol, TRC
Produktkod. 30010750
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010750 500 mg
30004536 100 mg
30004365 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD435633500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4453-82-1
Molekylformel C13H24O
Molekylvikt (g/mol) 196.32
Synonym Dicycloverine Impurity 7
LEDER OC(C1CCCCC1)C2CCCCC2
Specifikationer
Kemiskt namn eller material Dicyclohexylmethanol
Anteckningar om renhetsgrad GC
Kvantitet 500 mg
InChI formel InChI=1S/C13H24O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-14H,1-10H2
Formel vikt 196.1827
Procent renhet >95%