Läs mer
Dicyclomine Hydrochloride, TRC
Produktkod. 30136991
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136991 1 g
30132637 500 mg
17930455 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136991 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4393001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67-92-5
Molekylformel C19 H35 N O2 . Cl H
Molekylvikt (g/mol) 345.95
Synonym [1,1'-Bicyclohexyl]-1-carboxylic acid, 2-(diethylamino)ethyl ester, hydrochloride (9CI), [Bicyclohexyl]-1-carboxylic acid, 2-(diethylamino)ethyl ester hydrochloride, Atumin, Benacol, Bentomine, Bentyl hydrochloride, Di-Syntramine, Dicyclomine hydrochloride, Dicycloverin hydrochloride, Diocyl hydrochloride, Dyspas, Mamiesan, Merbentyl, Procyclomin, Wyovin hydrochloride, β-Diethylaminoethyl 1-cyclohexylcyclohexanecarboxylate hydrochloride
IUPAC-namn 2-(diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate;hydrochloride
LEDER Cl.CCN(CC)CCOC(=O)C1(CCCCC1)C2CCCCC2
Specifikationer
Kemiskt namn eller material Dicycloverine Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C19H35NO2.ClH/c1-3-20(4-2)15-16-22-18(21)19(13-9-6-10-14-19)17-11-7-5-8-12-17;/h17H,3-16H2,1-2H3;1H
Formel vikt 345.2435
Procent renhet >95%