Learn More
Dicyclopentenyl acrylate, Thermo Scientific Chemicals
Catalog Number p-7080229
Click to view available options
Quantity:
25 g
100 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
33791-58-1
C13H17O2
205.28
MFCD00171305
DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
33791-58-1
C13H17O2
205.28
MFCD00171305
DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate
Dicyclopentenyl acrylate is used in organic synthesis of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -40°C
Color Brown
Density 1.07 g/mL
Flash Point 131°C (268°F)
Quantity 25 g
Solubility Information Fully miscible in water.
Formula Weight 204.27
Physical Form Liquid
Chemical Name or Material Dicyclopentenyl acrylate