Läs mer
Tocris Bioscience™ DiD perchlorate
Lipophilic fluorescent reagent; shown to stain HSCs
Brand: Tocris Bioscience™ 5702/25
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
DiD perchlorate is a lipophilic fluorescent reagent; labels membranes and other hydrophobic structures. Shown to stain HSCs. Excitation/emission λ 644/665 nm.Specifikationer
| 2-[5-(1,3-Dihydro-3,3-dimethyl-1-octadecyl-2H-indol-2-ylidene)-1,3-pentadien-1-yl]-3,3-dimethyl-1-octadecyl-3H-indolium perchlorate | |
| 127274-91-3 | |
| 25 mg |