Läs mer
Didecyldimethylammonium chloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Produktkod. 30315150
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30315150 1 ml
1 options
Produktkod. 30315150 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA12588000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Didecyldimethylammonium chloride (DDAC-C10)
CAS 7173-51-5
Analyt- eller komponentnamn Didecyldimethylammonium chloride (DDAC-C10)
Frakt skick Room Temperature
Molekylformel C22 H48 N . Cl
Koncentration 100 μg/mL
Certifikat och efterlevnad ISO 17034
Synonym DDAC-C10, Didecyldimethylammonium-chloride
InChI formel InChI=1S/C22H48N.ClH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1
LEDER [Cl-].CCCCCCCCCC[N+](C)(C)CCCCCCCCCC
IUPAC-namn didecyl(dimethyl)azanium;chloride
Lösningsmedel eller matris Acetonitrile
Molekylvikt (g/mol) 362.08
Kvantitet 1 ml
Formel vikt 361.3475 g/mol