Läs mer
Dideiodo Amiodarone Hydrochloride, TRC
Produktkod. 30374729
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374729 100 mg
30372173 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD439425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95820-13-6
Molekylformel C25 H31 N O3 . Cl H
Molekylvikt (g/mol) 429.98
Synonym Methanone, (2-butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]phenyl]-, hydrochloride (1:1), Ketone, 2-butyl-3-benzofuranyl p-[2-(diethylamino)ethoxy]phenyl, hydrochloride (7CI), (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]phenyl]methanone Hydrochloride, Dideiodo Amiodarone hydrochloride
IUPAC-namn (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]phenyl]methanone;hydrochloride
LEDER Cl.CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)cc3
Specifikationer
Kemiskt namn eller material Dideiodo Amiodarone Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C25H31NO3.ClH/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;/h8-10,12-16H,4-7,11,17-18H2,1-3H3;1H
Formel vikt 429.2071