Läs mer
Didesmethyl Almotriptan, TRC
Produktkod. 30375745
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30375745 5 mg
1 options
Produktkod. 30375745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4408305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Didesmethyl Almotriptan
CAS 181178-24-5
Molekylformel C15H21N3O2S
Kvantitet 5 mg
Synonym 1-[[3-(2-Aminoethyl)-5-indolyl]methanesulfonyl]pyrrolidine, 1-[[[3-(2-Aminoethyl)-1H-indol-5-yl]methyl]sulfonyl]pyrrolidine, 5-[(1-Pyrrolidinylsulfonyl)methyl]-1H-indole-3-ethanamine, Almotriptan USP RC B, Almotriptan USP Related Compound B
InChI formel InChI=1S/C15H21N3O2S/c16-6-5-13-10-17-15-4-3-12(9-14(13)15)11-21(19,20)18-7-1-2-8-18/h3-4,9-10,17H,1-2,5-8,11,16H2
LEDER NCCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12
IUPAC-namn 2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]ethanamine
Molekylvikt (g/mol) 307.41
Formel vikt 307.1354