Läs mer
Didesmethyl Cariprazine, TRC
Produktkod. 30005535
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30005535 25 mg -
30009376 2.5 mg 2.5mg
30003927 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44166325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 839712-25-3
Molekylformel C19H28Cl2N4O
Molekylvikt (g/mol) 399.36
Synonym N-[trans-4-[2-[4-(2,3-Dichlorophenyl)-1-piperazinyl]ethyl]cyclohexyl]urea
LEDER ClC(C(Cl)=CC=C1)=C1N(CC2)CCN2CC[C@H]3CC[C@H](NC(N)=O)CC3
Specifikationer
Kemiskt namn eller material Didesmethyl Cariprazine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C19H28Cl2N4O/c20-16-2-1-3-17(18(16)21)25-12-10-24(11-13-25)9-8-14-4-6-15(7-5-14)23-19(22)26/h1-3,14-15H,4-13H2,(H3,22,23,26)
Formel vikt 398.164
Procent renhet >95%