missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didodecyl 3,3'-Thiodipropionate, TRC
Specifications
Specifications
| Kemiskt namn eller material | 3,3'-Thiodipropionic acid didodecyl ester |
| CAS | 123-28-4 |
| Molekylformel | C30 H58 O4 S |
| Kvantitet | 10 g |
| Synonym | 3,3'-Thiodipropionic Acid Didodecyl Ester, Didodecyl 3,3'-thiodipropionate, Propanoic acid, 3,3'-thiobis-, 1,1'-didodecyl ester (ACI), Propanoic acid, 3,3'-thiobis-, didodecyl ester (9CI), Propionic acid, 3,3'-thiodi-, didodecyl ester (6CI, 8CI), 3,3'-Thiodipropionic acid di(n-dodecyl) ester, Advastab 800, Antiox DLTP, Antiox L, Antioxidant 800, Antioxidant AS, Antioxidant DLDP, Antioxidant DLTDP, Antioxidant DLTP, Antioxidant LTDP, Antioxidant TPL, Arbestab DLTP, Bis(dodecyloxycarbonylethyl) sulfide, BNX DLTDP, Carstab DLTDP, Chinox DLTP, Cyanox LTDP, D 1, D 1 (antioxidant), Didodecyl 3,3'-thiodipropionate, Dilauryl 3,3'-thiodipropionate, Dilauryl thiodipropionate, Dilauryl β,β'-thiodipropionate, Dilauryl β-thiodipropionate, DLT, DLTDP, DLTP, DMPTP, Gsinox DLTP, Hostanox SE 1, Ipognox 89, Irgafos PS 800, Irganox DLTP, Irganox PS 800, Irganox PS 800FD, Irganox PS 800FL, KY 800, Lauryl 3,3'-thiodipropionate, Lusmit, Milban F, Naugard DLTDP, Neganox DLTP, Nocrac 400, Nonox DLTDP, NSC 65494, Plastanox LTDP, Plastanox LTDP Antioxidant, PS 800, PS 800FD, PS 800FL, Rasumitto, Rasumitto LG, Songnox DLTDP, Sonox DLTDP, Stabilizer DLT, Sumilizer TPL, Sumilizer TPL-R, TF 6693, Thanox DLTP, Thiobis(dodecyl propionate), Thiodipropionic acid didodecyl ester, Tinogard DA, TPL, TPL-R, Tyox B, Yoshinox DLTP |
| InChI formel | InChI=1S/C30H58O4S/c1-3-5-7-9-11-13-15-17-19-21-25-33-29(31)23-27-35-28-24-30(32)34-26-22-20-18-16-14-12-10-8-6-4-2/h3-28H2,1-2H3 |
| LEDER | CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC |
| IUPAC-namn | dodecyl 3-(3-dodecoxy-3-oxopropyl)sulfanylpropanoate |
| Molekylvikt (g/mol) | 514.84 |
| Formel vikt | 514.4056 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?